Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000284.3 | NP_000275.1 | 11 | 1338 | ||
NM_001173454.1 | NP_001166925.1 | 12 | 1452 | |||
NM_001173455.1 | NP_001166926.1 | 11 | 1359 | |||
NM_001173456.1 | NP_001166927.1 | 10 | 1245 | |||
XM_017029574.1 | XP_016885063.1 | 11 | 1346 | |||
GenBank mRNA | AK129887.1 | - | 11 | 1273 | ||
AK222740.1 | - | 11 | 1269 | |||
AK222988.1 | - | 11 | 1303 | |||
AK296341.1 | - | 11 | 1325 | |||
AK296457.1 | - | 10 | 1159 | |||
BC002406.1 | - | 11 | 1250 | 2961286 | ||
EF576990.1 | - | 8 | 807 | |||
EF590116.1 | - | 5 | 988 | |||
EF590117.1 | - | 1370 | ||||
J03503.1 | - | 11 | 1265 | |||
J03575.1 | - | 11 | 1238 | |||
L13318.1 | - | 11 | 1298 | |||
L48690.1 | - | 11 | 1298 | |||
M24848.1 | - | 11 | 1236 | |||
X52709.1 | - | 11 | 1247 | |||
X52710.1 | - | 11 | 1249 |