Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAPDH SIRNA Assays ›
          Gene Symbol:
          GAPDH GAPDHP44 GAPDHP62
          Gene Name:
          glyceraldehyde-3-phosphate dehydrogenase
          glyceraldehyde 3 phosphate dehydrogenase pseudogene 44
          glyceraldehyde 3 phosphate dehydrogenase pseudogene 62
          Gene Aliases:
          G3PD, GAPD, HEL-S-162eP
          GAPDH
          GAPDHL7, GAPDL7
          Chromosome Location:
          Chr.12: 6534405 - 6538375 on Build GRCh38
          Chromosome Location:
          Chr.12: 62755153 - 62756433 on Build GRCh38
          Chromosome Location:
          Chr.8: 100550540 - 100551790 on Build GRCh38
          UniGene ID:
          Hs.544577
          Hs.732374
          Hs.700353
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 212721

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GAPDH
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256799.2 NP_001243728.1 5 576
          NM_001289745.1 NP_001276674.1 6 634
          NM_001289746.1 NP_001276675.1 5 528
          NM_002046.5 NP_002037.2 6 542
          GenBank mRNA AB062273.1 - 6 406
          AF261085.1 - 6 457
          AK026525.1 - 6 479
          AK299448.1 - 6 427
          AK308198.1 - 6 430
          AY007133.1 - 6 406
          AY633612.1 - 5 378
          BC001601.1 - 6 408 3543589
          BC004109.2 - 6 406 3678259
          BC009081.1 - 6 415 3869809
          BC013310.2 - 6 414 3538383
          BC020308.1 - 5 392
          BC023632.2 - 5 378 4842418
          BC025925.1 - 6 411 2989348
          BC026907.1 - 5 392 4454083
          BC029340.1 - 6 400
          BC029618.1 - 6 408 3876535
          BC083511.1 - 6 406 3536969
          BG724119.1 - 5 559
          BI463134.1 - 5 558
          BM763361.1 - 6 519
          BT006893.1 - 5 354
          BU155402.1 - 6 579
          CR407671.1 - 5 354
          DQ403057.1 - 4 279
          EF036498.1 - 2 92
          EU668321.1 - 6 456
          J02642.1 - 5 355
          M17851.1 - 5 387
          M33197.1 - 6 414
          X01677.1 - 5 414
          X53778.1 - 6 410
          Gene Symbol: GAPDHP44
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA JN038570.1 - 1 261
          Gene Symbol: GAPDHP62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC040702.1 - 2 492

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001289726, NM_008084, BU504528, BC083079, AK168217, and others.

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          gluconeogenesis
          regulation of macroautophagy
          negative regulation of translation
          peptidyl-cysteine S-trans-nitrosylation
          protein stabilization
          neuron apoptotic process
          canonical glycolysis
          cellular response to interferon-gamma
          glyceraldehyde-3-phosphate dehydrogenase (NAD+) (phosphorylating) activity
          protein binding
          microtubule binding
          peptidyl-cysteine S-nitrosylase activity
          identical protein binding
          NADP binding
          NAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0