Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271957.1 | NP_001258886.1 | 2 | 516 | ||
NM_001271958.1 | NP_001258887.1 | 2 | 353 | |||
NM_001271959.1 | NP_001258888.1 | 2 | 522 | |||
NM_001271960.1 | NP_001258889.1 | 3 | 414 | |||
NM_001271961.1 | NP_001258890.1 | 2 | 353 | |||
NM_014437.4 | NP_055252.2 | 3 | 660 | |||
GenBank mRNA | AF132942.1 | - | 1 | 210 | ||
AF151829.1 | - | 2 | 231 | |||
AJ271671.1 | - | 2 | 261 | |||
AK074943.1 | - | 2 | 470 | |||
AK293389.1 | - | 2 | 261 | |||
AK297983.1 | - | 3 | 228 | |||
AK300244.1 | - | 3 | 220 | |||
BC002563.2 | - | 2 | 235 | 3138575 | ||
BC003152.2 | - | 1 | 207 | 2958651 | ||
BC007886.2 | - | 2 | 232 | 4122086 | ||
BC014303.2 | - | 3 | 640 | 3942586 | ||
CR457217.1 | - | 1 | 174 | |||
DA052424.1 | - | 2 | 354 | |||
DA354647.1 | - | 2 | 498 | |||
DA711655.1 | - | 2 | 363 | |||
DA818910.1 | - | 2 | 340 | |||
DB001496.1 | - | 3 | 414 |