Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYNJ1 SIRNA Assays ›
          Gene Symbol:
          SYNJ1
          Gene Name:
          synaptojanin 1
          Gene Aliases:
          INPP5G, PARK20
          Chromosome Location:
          Chr.21: 32628759 - 32728128 on Build GRCh38
          UniGene ID:
          Hs.473632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 212747

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYNJ1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001160302.1 NP_001153774.1 12 1555
          NM_001160306.1 NP_001153778.1 12 1564
          NM_003895.3 NP_003886.3 12 1547
          NM_203446.2 NP_982271.2 12 1547
          XM_017028494.1 XP_016883983.1 12 1537
          XM_017028495.1 XP_016883984.1 12 1571
          XM_017028496.1 XP_016883985.1 12 1537
          XM_017028497.1 XP_016883986.1 12 1635
          XM_017028498.1 XP_016883987.1 12 1537
          XM_017028499.1 XP_016883988.1 12 1571
          XM_017028500.1 XP_016883989.1 12 1537
          XM_017028501.1 XP_016883990.1 12 1537
          XM_017028502.1 XP_016883991.1 12 1537
          XM_017028503.1 XP_016883992.1 12 1537
          XM_017028504.1 XP_016883993.1 12 1537
          XM_017028505.1 XP_016883994.1 12 1571
          GenBank mRNA AB020717.2 - 12 1539
          AF009039.1 - 12 1473
          AF009040.1 - 12 1473
          AK307762.1 - 12 1537
          BC019630.1 - 12 1546
          BC098395.1 - 12 1525
          BC136603.1 - 11 1460 9020593

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          neurotransmitter transport
          learning
          synaptic vesicle priming
          synaptic vesicle uncoating
          phosphatidylinositol-3-phosphate biosynthetic process
          inositol phosphate metabolic process
          phosphatidylinositol metabolic process
          inositol phosphate dephosphorylation
          phosphatidylinositol dephosphorylation
          synaptic vesicle endocytosis
          synaptic vesicle transport
          positive regulation of endosome organization
          nucleotide binding
          RNA binding
          phosphatidylinositol-3-phosphatase activity
          phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
          phosphatidylinositol phosphate 5-phosphatase activity
          phosphatidylinositol phosphate 4-phosphatase activity
          phosphatidylinositol-4-phosphate phosphatase activity
          phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
          phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
          inositol-1,4,5-trisphosphate 5-phosphatase activity
          inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0