Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNE1 SIRNA Assays ›
          Gene Symbol:
          KCNE1 KCNE1B
          Gene Name:
          potassium voltage-gated channel subfamily E regulatory subunit 1
          potassium voltage-gated channel subfamily E regulatory subunit 1B
          Gene Aliases:
          ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK
          -
          Chromosome Location:
          Chr.21: 34446688 - 34512275 on Build GRCh38
          Chromosome Location:
          Chr.21: 7816677 - 7829632 on Build GRCh38
          UniGene ID:
          Hs.121495
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 212857

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KCNE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000219.5 NP_000210.2 4 632
          NM_001127668.3 NP_001121140.1 3 244
          NM_001127669.3 NP_001121141.1 3 216
          NM_001127670.3 NP_001121142.1 2 481
          NM_001270402.2 NP_001257331.1 3 407
          NM_001270403.2 NP_001257332.1 3 355
          NM_001270404.2 NP_001257333.1 2 296
          NM_001270405.2 NP_001257334.1 3 208
          XM_017028342.1 XP_016883831.1 2 1984
          GenBank mRNA AK311384.1 - 3, 4 199
          AK313543.1 - 3 239
          AY789479.1 - 3 171
          AY789480.1 - 2 413
          BC036452.1 - 3 218 5266786
          BC046224.1 - 4 632 6198685
          DA418828.1 - 3 408
          DA806205.1 - 3 208
          DB089177.1 - 2 437
          DB214219.1 - 2 256
          DB223354.1 - 2 287
          DB230009.1 - 3 330
          DB233067.1 - 3 397
          EU008568.1 - 3 235
          EU008569.1 - 3 207
          EU008570.1 - 2 482
          L28168.1 - 1 25
          Gene Symbol: KCNE1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006723946.2 XP_006724009.1 2 437
          XM_006723949.2 XP_006724012.1 3 284

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          protein N-linked glycosylation
          protein O-linked glycosylation
          sensory perception of sound
          regulation of ventricular cardiac muscle cell membrane repolarization
          cardiac conduction
          cellular response to cAMP
          potassium ion export
          potassium ion transmembrane transport
          cardiac muscle cell action potential involved in contraction
          ventricular cardiac muscle cell action potential
          membrane repolarization
          membrane repolarization during action potential
          membrane repolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          negative regulation of protein targeting to membrane
          membrane repolarization during ventricular cardiac muscle cell action potential
          regulation of potassium ion transmembrane transport
          positive regulation of potassium ion transmembrane transport
          regulation of delayed rectifier potassium channel activity
          negative regulation of delayed rectifier potassium channel activity
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          protein binding
          potassium channel regulator activity
          telethonin binding
          voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0