Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253829.1 | NP_001240758.1 | 2 | 375 | ||
NM_001253830.1 | NP_001240759.1 | 3 | 436 | |||
NM_152422.4 | NP_689635.3 | 2 | 369 | |||
NM_177995.2 | NP_818931.1 | 3 | 451 | |||
XM_005251711.4 | XP_005251768.1 | 3 | 2393 | |||
XM_011518240.2 | XP_011516542.1 | 3 | 201 | |||
XM_011518241.2 | XP_011516543.1 | 3 | 155 | |||
XM_017014280.1 | XP_016869769.1 | 4 | 260 | |||
XM_017014281.1 | XP_016869770.1 | 3 | 179 | |||
GenBank mRNA | AK124912.1 | - | 2 | 255 | ||
AK289692.1 | - | 2 | 326 | |||
AK310403.1 | - | 2 | 333 | |||
AY171233.1 | - | 3 | 456 | |||
BC067120.1 | - | 3 | 436 | 5539182 | ||
BI460503.1 | - | 3 | 457 | |||
DA218576.1 | - | 2 | 369 |