Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CADM1 SIRNA Assays ›
          Gene Symbol:
          CADM1
          Gene Name:
          cell adhesion molecule 1
          Gene Aliases:
          BL2, IGSF4, IGSF4A, NECL2, Necl-2, RA175, ST17, SYNCAM, TSLC1, sTSLC-1, sgIGSF, synCAM1
          Chromosome Location:
          Chr.11: 115173625 - 115504523 on Build GRCh38
          UniGene ID:
          Hs.370510
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 21305

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CADM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098517.1 NP_001091987.1 2 379
          NM_001301043.1 NP_001287972.1 2 379
          NM_001301044.1 NP_001287973.1 2 379
          NM_001301045.1 NP_001287974.1 2 379
          NM_014333.3 NP_055148.3 2 379
          XM_005271494.2 XP_005271551.1 2 272
          XM_017017457.1 XP_016872946.1 2 338
          XM_017017458.1 XP_016872947.1 2 145
          XM_017017459.1 XP_016872948.1 3 2332
          XM_017017460.1 XP_016872949.1 3 2332
          XM_017017461.1 XP_016872950.1 3 346
          GenBank mRNA AB094146.1 - 2 250
          AF132811.1 - 2 253
          AK075502.1 - 2 269
          AK315245.1 - 2 312
          AY358334.1 - 2 265
          BC035930.1 - 2 271
          BC047021.1 - 2 379 5248100
          BC125102.1 - 2 254
          BC125103.1 - 2 253 40118330
          BI915307.1 - 2 379
          CR457157.1 - 2 250
          HE586496.1 - 2 253
          HE586497.1 - 2 253
          HE586498.1 - 2 253
          HE586499.1 - 2 253
          HE586500.1 - 3 339
          HE586503.1 - 3 281
          KJ534791.1 - 2 250
          KJ534792.1 - 2 250
          KJ534793.1 - 2 250
          KJ534794.1 - 2 250
          KJ534795.1 - 2 250
          KJ534796.1 - 2 250
          KJ534797.1 - 2 250

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          T cell mediated cytotoxicity
          apoptotic process
          homophilic cell adhesion via plasma membrane adhesion molecules
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          multicellular organism development
          spermatogenesis
          cell recognition
          cell differentiation
          adherens junction organization
          susceptibility to natural killer cell mediated cytotoxicity
          positive regulation of natural killer cell mediated cytotoxicity
          positive regulation of cytokine secretion
          activated T cell proliferation
          detection of stimulus
          receptor activity
          receptor binding
          protein binding
          PDZ domain binding
          protein homodimerization activity
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0