Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D22A SIRNA Assays ›
Gene Symbol:
TBC1D22A
Gene Name:
TBC1 domain family member 22A
Gene Aliases:
C22orf4, HSC79E021
Chromosome Location:
Chr.22: 46762617 - 47224975 on Build GRCh38
UniGene ID:
Hs.435044
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21314
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D22A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284303.1
NP_001271232.1
2, 3
583
NM_001284304.1
NP_001271233.1
5, 6
954
NM_001284305.1
NP_001271234.1
4, 5
894
NM_014346.3
NP_055161.1
4, 5
817
NR_104292.1
5, 6
1260
XM_005261496.1
XP_005261553.1
4, 5
813
XM_011530099.1
XP_011528401.1
4, 5
813
XM_011530100.2
XP_011528402.1
4, 5
813
XM_011530101.2
XP_011528403.1
4, 5
813
XM_011530102.2
XP_011528404.1
4, 5
813
XM_017028742.1
XP_016884231.1
4, 5
813
XM_017028743.1
XP_016884232.1
4, 5
813
XM_017028744.1
XP_016884233.1
5, 6
805
XM_017028745.1
XP_016884234.1
3, 4
636
XM_017028746.1
XP_016884235.1
4, 5
813
XM_017028747.1
XP_016884236.1
4, 5
813
GenBank mRNA
AB449908.1
-
4, 5
647
AK055591.1
-
4, 5
805
AK125705.1
-
5, 6
1260
AK127653.1
-
4, 5
894
AK301445.1
-
4, 5
453
AK308039.1
-
5, 6
882
AK315779.1
-
4, 5
779
AL096779.1
-
11
BC001292.1
-
4, 5
750
3457393
BC002743.2
-
2, 3
519
3627634
BC020976.1
-
4, 5
752
3884347
BC029897.1
-
4, 5
782
5173045
CR456412.1
-
4, 5
668
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
regulation of vesicle fusion
activation of GTPase activity
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
protein homodimerization activity
14-3-3 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0