Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014512.1 | NP_055327.1 | 3 | 101 | ||
GenBank mRNA | AF022046.1 | - | 3 | 101 | ||
AF285437.1 | - | 2 | 48 | |||
AM999888.1 | - | 3 | 96 | |||
X89892.1 | - | 3 | 108 | |||
X98858.1 | - | 3 | 88 |
Gene Symbol: 1060P11.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ630586.1 | - | 4 | 374 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014218.2 | NP_055033.2 | 3 | 126 | ||
XM_011526939.2 | XP_011525241.1 | 3 | 731 | |||
XM_017026782.1 | XP_016882271.1 | 3 | 104 | |||
XM_017026783.1 | XP_016882272.1 | 3 | 731 | |||
GenBank mRNA | AF022045.1 | - | 3 | 95 | ||
AF285431.1 | - | 3 | 121 | |||
AF285432.1 | - | 2 | 48 | |||
AY366237.1 | - | 3 | 124 | |||
AY366238.1 | - | 3 | 70 | |||
AY366239.1 | - | 3 | 118 | |||
AY366240.1 | - | 3 | 126 | |||
AY366241.1 | - | 3 | 125 | |||
AY388471.1 | - | 3 | 118 | |||
AY789055.1 | - | 3 | 121 | |||
AY789056.1 | - | 3 | 88 | |||
BC069344.1 | - | 3 | 121 | 7262296 | ||
KJ699235.1 | - | 3 | 361 | |||
L41267.1 | - | 3 | 120 | |||
U24076.1 | - | 3 | 121 | |||
U24078.1 | - | 3 | 128 | |||
U96189.1 | - | 1 | 25 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 117 | ||
GenBank mRNA | AF285434.1 | - | 2 | 48 | ||
AY366242.1 | - | 3 | 117 | |||
KP260924.1 | - | 3 | 84 | |||
L76669.1 | - | 3 | 99 | |||
U24075.1 | - | 3 | 102 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015868.2 | NP_056952.2 | 3 | 125 | ||
GenBank mRNA | AF135556.1 | - | 1 | 25 | ||
AF135566.1 | - | 1 | 25 | |||
AF135567.1 | - | 1 | 25 | |||
AF285435.1 | - | 2 | 48 | |||
AY789057.1 | - | 3 | 88 | |||
BC032422.1 | - | 3 | 101 | 5220339 | ||
BC050730.1 | - | 3 | 119 | 5219336 | ||
BC110613.1 | - | 3 | 102 | 40031619 | ||
FN395076.1 | - | 3 | 88 | |||
FN552550.1 | - | 3 | 70 | |||
L41268.1 | - | 3 | 125 | |||
L76662.1 | - | 3 | 99 | |||
L76663.1 | - | 3 | 99 | |||
U24074.1 | - | 3 | 109 | |||
U50546.1 | - | 3 | 99 | |||
U96190.1 | - | 1 | 25 |
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 123 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 123 | |||
NM_012312.4 | NP_036444.1 | 3 | 123 | |||
GenBank mRNA | AF135565.1 | - | 1 | 25 | ||
AF285438.1 | - | 2 | 48 | |||
AJ002102.1 | - | 3 | 88 | |||
AJ002103.1 | - | 3 | 88 | |||
AJ002104.1 | - | 3 | 88 | |||
AJ002105.1 | - | 3 | 88 | |||
AM409094.1 | - | 3 | 89 | |||
AY366243.1 | - | 3 | 98 | |||
AY366244.1 | - | 3 | 87 | |||
BC108917.1 | - | 3 | 90 | 40017299 | ||
BC119631.1 | - | 3 | 107 | 40115568 | ||
BC119632.1 | - | 3 | 107 | 40115570 | ||
BC125146.1 | - | 2 | 72 | |||
BC126856.1 | - | 3 | 90 | 40017301 | ||
FR750994.1 | - | 3 | 123 | |||
L41347.1 | - | 3 | 107 | |||
L76667.1 | - | 3 | 99 | |||
U24079.1 | - | 3 | 86 | |||
X89893.1 | - | 3 | 88 |
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3 | 99 | ||
GenBank mRNA | AF022047.1 | - | 3 | 92 | ||
AM851002.1 | - | 3 | 124 | |||
AY789063.2 | - | 2 | 72 | |||
L76670.1 | - | 3 | 99 | |||
X97231.1 | - | 3 | 147 |
Gene Symbol: KIR2DS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014513.2 | NP_055328.2 | 3 | 122 | ||
GenBank mRNA | AF208054.1 | - | 3 | 122 | ||
AF272389.1 | - | 3 | 72 | |||
AY366247.1 | - | 3 | 146 | |||
AY366248.1 | - | 3 | 146 | |||
L76672.1 | - | 3 | 99 |
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 4 | 436 | ||
NM_013289.2 | NP_037421.2 | 4 | 436 | |||
GenBank mRNA | AF022049.1 | - | 4 | 373 | ||
AF103010.1 | - | 4 | 373 | |||
AF103011.1 | - | 4 | 373 | |||
AF103012.1 | - | 4 | 373 | |||
AF103013.1 | - | 4 | 373 | |||
AF208688.1 | - | 2 | 310 | |||
AF262968.1 | - | 2 | 292 | |||
AF262969.1 | - | 2 | 292 | |||
AF262970.1 | - | 2 | 292 | |||
AF262971.1 | - | 2 | 292 | |||
AF262972.1 | - | 2 | 292 | |||
AF262973.1 | - | 4 | 406 | |||
AF262974.1 | - | 4 | 406 | |||
AJ417556.1 | - | 2 | 322 | |||
AJ417557.1 | - | 2 | 322 | |||
AY366236.1 | - | 4 | 411 | |||
AY366249.1 | - | 4 | 373 | |||
AY366250.1 | - | 4 | 399 | |||
AY366251.1 | - | 4 | 373 | |||
AY366252.1 | - | 4 | 373 | |||
AY760050.1 | - | 4 | 373 | |||
AY789065.1 | - | 4 | 388 | |||
AY789066.1 | - | 4 | 373 | |||
AY789072.1 | - | 4 | 373 | |||
BC028206.1 | - | 4 | 400 | 5215674 | ||
BC101540.1 | - | 4 | 392 | 8069046 | ||
BC101542.1 | - | 4 | 392 | 8069048 | ||
DQ178659.1 | - | 4 | 373 | |||
EF462885.1 | - | 2 | 303 | |||
EF472674.1 | - | 2 | 303 | |||
EF472675.1 | - | 2 | 303 | |||
EF472676.1 | - | 2 | 303 | |||
EF472677.1 | - | 2 | 303 | |||
EF472678.1 | - | 2 | 303 | |||
EF472679.1 | - | 2 | 303 | |||
L41269.1 | - | 4 | 392 | |||
L76664.1 | - | 3 | 99 | |||
U30273.1 | - | 4 | 375 | |||
U30274.1 | - | 4 | 436 | |||
U31416.1 | - | 4 | 398 | |||
U33328.1 | - | 4 | 398 | |||
X94262.1 | - | 4 | 374 |
Gene Symbol: KIR3DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242867.1 | NP_001229796.1 | 4 | 406 | ||
NM_006737.3 | NP_006728.2 | 4 | 406 | |||
XM_011526940.2 | XP_011525242.1 | 4 | 530 | |||
XM_017026784.1 | XP_016882273.1 | 4 | 530 | |||
GenBank mRNA | AF208683.1 | - | 2 | 310 | ||
AF208684.1 | - | 2 | 310 | |||
AF208685.1 | - | 2 | 310 | |||
AF208686.1 | - | 2 | 310 | |||
AF208690.1 | - | 2 | 310 | |||
AF262965.1 | - | 2 | 292 | |||
AF262966.1 | - | 2 | 292 | |||
AF262967.1 | - | 4 | 406 | |||
AF263617.1 | - | 4 | 406 | |||
AJ276125.1 | - | 4 | 374 | |||
AW951147.1 | - | 1 | 31 | |||
AY059418.1 | - | 2 | 304 | |||
AY059419.1 | - | 2 | 304 | |||
AY059420.1 | - | 2 | 304 | |||
AY366256.2 | - | 4 | 385 | |||
AY789067.1 | - | 4 | 373 | |||
AY789068.1 | - | 4 | 373 | |||
AY789069.1 | - | 3 | 343 | |||
AY789070.1 | - | 4 | 373 | |||
AY789071.1 | - | 3 | 343 | |||
BC105678.1 | - | 4 | 398 | 40004817 | ||
BC107951.1 | - | 4 | 398 | 40004820 | ||
DQ256504.1 | - | 2 | 305 | |||
EF582384.1 | - | 4 | 373 | |||
L41270.1 | - | 4 | 392 | |||
L76665.1 | - | 4 | 384 | |||
L76666.1 | - | 4 | 384 | |||
U30272.1 | - | 4 | 375 | |||
X93595.1 | - | 4 | 403 | |||
X93596.1 | - | 4 | 373 | |||
X94373.1 | - | 4 | 374 | |||
X94374.1 | - | 4 | 373 |
Gene Symbol: KIR3DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153443.4 | NP_703144.3 | 4 | 423 | ||
GenBank mRNA | AF352324.1 | - | 4 | 391 | ||
AJ938052.1 | - | 4 | 378 | |||
AJ938053.1 | - | 4 | 378 | |||
AJ938054.1 | - | 4 | 378 | |||
AJ938055.1 | - | 4 | 378 | |||
AJ938056.1 | - | 4 | 378 | |||
AJ938057.1 | - | 4 | 378 | |||
AJ938058.1 | - | 4 | 378 | |||
AJ938059.1 | - | 4 | 378 | |||
AJ938060.1 | - | 4 | 378 | |||
AJ938061.1 | - | 4 | 378 | |||
AJ938062.1 | - | 4 | 378 | |||
AJ938063.1 | - | 4 | 378 | |||
AJ938064.1 | - | 4 | 378 | |||
AJ938065.1 | - | 4 | 378 | |||
AJ938066.1 | - | 4 | 378 | |||
BC136283.1 | - | 4 | 423 | 9020270 | ||
BC143802.1 | - | 4 | 423 | 9052316 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 4 | 410 | ||
NM_001282170.1 | NP_001269099.1 | 3 | 125 | |||
NM_001282171.1 | NP_001269100.1 | 2 | 89 | |||
GenBank mRNA | AF022044.1 | - | 4 | 386 | ||
AJ417558.3 | - | 2 | 322 | |||
AK301140.1 | - | 4 | 410 | |||
BC098107.1 | - | 2 | 63 | 40026977 | ||
BC098288.1 | - | 4 | 384 | 40026980 | ||
BC105679.1 | - | 4 | 384 | 40004313 | ||
DC390157.1 | - | 4 | 410 | |||
EF370406.1 | - | 3 | 334 | |||
EF582383.1 | - | 4 | 373 | |||
L76661.1 | - | 4 | 384 | |||
U73396.1 | - | 4 | 373 | |||
X97232.1 | - | 3 | 88 | |||
X97233.1 | - | 4 | 373 |