Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012313.1 | NP_036445.1 | 3 | 291 | ||
GenBank mRNA | AF022047.1 | - | 3 | 284 | ||
AM851002.1 | - | 3 | 316 | |||
AY789063.2 | - | 2 | 264 | |||
L76670.1 | - | 3 | 291 | |||
X97231.1 | - | 3 | 339 |
Gene Symbol: KIR2DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X89892.1 | - | 3 | 300 |