Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRE SIRNA Assays ›
Gene Symbol:
PTPRE
Gene Name:
protein tyrosine phosphatase, receptor type E
Gene Aliases:
HPTPE, PTPE, R-PTP-EPSILON
Chromosome Location:
Chr.10: 127907061 - 128085855 on Build GRCh38
UniGene ID:
Hs.127022
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2132
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001316676.1
NP_001303605.1
7
780
NM_001316677.1
NP_001303606.1
5
453
NM_001323354.1
NP_001310283.1
5
2173
NM_001323355.1
NP_001310284.1
5
586
NM_001323356.1
NP_001310285.1
5
580
NM_001323357.1
NP_001310286.1
3
256
NM_006504.5
NP_006495.1
6
609
NM_130435.4
NP_569119.1
3
256
XM_005252691.2
XP_005252748.1
5
453
XM_011539994.2
XP_011538296.1
5
571
XM_011539995.1
XP_011538297.1
5
571
XM_011539996.1
XP_011538298.1
5
571
XM_011539998.1
XP_011538300.1
5
571
XM_017016467.1
XP_016871956.1
5
571
XM_017016468.1
XP_016871957.1
5
360
XM_017016469.1
XP_016871958.1
5
571
GenBank mRNA
AF406557.1
-
6
544
AJ315969.1
-
3
165
AJ430580.1
-
3
174
AK056111.1
-
5
2173
AK290414.1
-
3
192
AK291828.1
-
5
453
AK299498.1
-
5
422
AK300613.1
-
4
346
BC050062.1
-
7
688
5274802
BE246451.1
-
3
237
BI559814.1
-
6
616
BI910569.1
-
3
219
BU159075.1
-
3
218
BX648180.1
-
3
262
DA470230.1
-
3
256
X54134.1
-
6
381
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
transmembrane receptor protein tyrosine phosphatase signaling pathway
regulation of mast cell activation
peptidyl-tyrosine dephosphorylation
negative regulation of insulin receptor signaling pathway
transmembrane receptor protein tyrosine phosphatase activity
protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-gskvx:80/100.66.75.242:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0