Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291695.1 | NP_001278624.1 | 3 | 197 | ||
NM_001291701.1 | NP_001278630.1 | 3 | 197 | |||
NM_012312.4 | NP_036444.1 | 3 | 197 | |||
GenBank mRNA | AF285438.1 | - | 2 | 122 | ||
AJ002102.1 | - | 3 | 162 | |||
AJ002103.1 | - | 3 | 162 | |||
AJ002104.1 | - | 3 | 162 | |||
AJ002105.1 | - | 3 | 162 | |||
AM409094.1 | - | 3 | 163 | |||
AY366243.1 | - | 3 | 172 | |||
AY366244.1 | - | 3 | 161 | |||
BC108917.1 | - | 3 | 164 | 40017299 | ||
BC119631.1 | - | 3 | 181 | 40115568 | ||
BC119632.1 | - | 3 | 181 | 40115570 | ||
BC125146.1 | - | 2 | 146 | |||
BC126856.1 | - | 3 | 164 | 40017301 | ||
FR750994.1 | - | 3 | 197 | |||
L41347.1 | - | 3 | 181 | |||
L76667.1 | - | 3 | 173 | |||
U24079.1 | - | 3 | 160 | |||
X89893.1 | - | 3 | 162 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 3 | 191 | ||
GenBank mRNA | AF285434.1 | - | 2 | 122 | ||
AY366242.1 | - | 3 | 191 | |||
KP260924.1 | - | 3 | 158 | |||
L76669.1 | - | 3 | 173 | |||
U24075.1 | - | 3 | 176 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022048.1 | - | 3 | 162 | ||
FN552550.1 | - | 3 | 144 | |||
U73395.1 | - | 3 | 183 |