Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000075.3 | NP_000066.1 | 5, 6 | 918 | ||
GenBank mRNA | AK297901.1 | - | 4, 5 | 661 | ||
AK313701.1 | - | 5, 6 | 742 | |||
BC003644.2 | - | 5, 6 | 765 | 3532812 | ||
BC005864.2 | - | 5, 6 | 829 | 2961345 | ||
BC010153.2 | - | 5, 6 | 767 | 3531300 | ||
BC015669.2 | - | 3, 5 | 451 | |||
CR407668.1 | - | 4, 5 | 626 | |||
CR542247.1 | - | 4, 5 | 626 | |||
M14505.1 | - | 5, 6 | 853 | |||
U79269.1 | - | 5, 6 | 713 | |||
Z48970.1 | - | 5, 6 | 760 |