Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBE3A SIRNA Assays ›
          Gene Symbol:
          UBE3A
          Gene Name:
          ubiquitin protein ligase E3A
          Gene Aliases:
          ANCR, AS, E6-AP, EPVE6AP, HPVE6A
          Chromosome Location:
          Chr.15: 25337234 - 25439086 on Build GRCh38
          UniGene ID:
          Hs.598862
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 213354

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBE3A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000462.3 NP_000453.2 7 1339
          NM_130838.1 NP_570853.1 3 554
          NM_130839.2 NP_570854.1 6 1274
          XM_005268267.4 XP_005268324.1 8 1641
          XM_005268268.4 XP_005268325.1 7 1528
          XM_005268269.4 XP_005268326.1 8 1372
          XM_005268270.4 XP_005268327.1 7 1329
          XM_005268271.4 XP_005268328.1 4 1148
          XM_006720675.3 XP_006720738.1 8 1593
          XM_006720676.3 XP_006720739.1 5 838
          XM_011521994.2 XP_011520296.1 8 1530
          XM_011521995.2 XP_011520297.1 8 1204
          XM_017022544.1 XP_016878033.1 9 1566
          XM_017022545.1 XP_016878034.1 8 1525
          XM_017022546.1 XP_016878035.1 8 1433
          XM_017022547.1 XP_016878036.1 7 1465
          XM_017022548.1 XP_016878037.1 7 1460
          XM_017022549.1 XP_016878038.1 7 1267
          XM_017022550.1 XP_016878039.1 7 1419
          XM_017022551.1 XP_016878040.1 8 1470
          XM_017022552.1 XP_016878041.1 8 1160
          XM_017022553.1 XP_016878042.1 8 1475
          XM_017022554.1 XP_016878043.1 4 1479
          XM_017022555.1 XP_016878044.1 6 1248
          XM_017022556.1 XP_016878045.1 7 1397
          XM_017022557.1 XP_016878046.1 6 1332
          XM_017022558.1 XP_016878047.1 7 1528
          XM_017022559.1 XP_016878048.1 4 1148
          GenBank mRNA AB194232.1 - 5 650
          AF273050.1 - 4 609
          AK056239.1 - 6 1242
          AK130491.1 - 2 283
          AK292514.1 - 6 884
          BC002582.2 - 6 1227 3160225
          L07557.1 - 3 549
          U84404.1 - 8 1096
          X98031.1 - 4 579
          X98032.1 - 2 510
          X98033.1 - 4 570

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191837, XM_008759399, XM_006229313, XM_006229310, XM_008759400, and others.
          This siRNA also targets the following transcripts in Mouse: NM_011668, NM_173010, NM_001033962, AK083795, AK029133, and others.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          proteolysis
          ubiquitin-dependent protein catabolic process
          brain development
          positive regulation of phosphatidylinositol 3-kinase signaling
          viral process
          androgen receptor signaling pathway
          positive regulation of protein ubiquitination
          sperm entry
          regulation of circadian rhythm
          positive regulation of transcription from RNA polymerase II promoter
          protein autoubiquitination
          prostate gland growth
          protein K48-linked ubiquitination
          regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          transcription coactivator activity
          ubiquitin-protein transferase activity
          protein binding
          ligase activity
          metal ion binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0