Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012270.1 | NP_001012270.1 | 3 | 1070 | ||
NM_001012271.1 | NP_001012271.1 | 5 | 1257 | |||
NM_001168.2 | NP_001159.2 | 4 | 1188 | |||
GenBank mRNA | AF077350.1 | - | 4 | 1091 | ||
AK223428.1 | - | 4 | 1134 | |||
AK301793.1 | - | 5 | 1306 | |||
BC000784.2 | - | 3 | 989 | |||
BC008718.2 | - | 4 | 1100 | 2961114 | ||
BC034148.1 | - | 4 | 1105 | 4656567 | ||
BC065497.1 | - | 4 | 1080 | 5394399 |