Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014394.2 | NP_055209.2 | 4, 5 | 548 | ||
GenBank mRNA | AB009685.1 | - | 3, 4 | 344 | ||
AF060923.1 | - | 4, 5 | 422 | |||
AF078863.1 | - | 4, 5 | 473 | |||
AF131820.1 | - | 4, 5 | 475 | |||
AF161400.1 | - | 4, 5 | 457 | |||
AK098770.1 | - | 5, 6 | 361 | |||
AK130094.1 | - | 4, 5 | 451 | |||
AK130097.1 | - | 4, 5 | 416 | |||
AK225318.1 | - | 4, 5 | 425 | |||
AK292864.1 | - | 4, 5 | 416 | |||
AK298332.1 | - | 4, 5 | 430 | |||
AK299937.1 | - | 3, 4 | 454 | |||
AY358824.1 | - | 4, 5 | 423 | |||
BC010354.1 | - | 4, 5 | 419 | 4093037 |