siRNA Details
Gene Symbol: CDK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001164410.2 | NP_001157882.1 | 8 | 659 | ||
NM_004935.3 | NP_004926.1 | 9 | 755 | |||
GenBank mRNA | AK026533.1 | - | 9 | 676 | ||
AY049778.1 | - | 9 | 638 | |||
AY927560.1 | - | LIT3563 AY927560 |
4 | 622 | ||
BC005115.1 | - | 9 | 663 | 3537202 | ||
BT006680.1 | - | 9 | 638 | |||
BU527089.1 | - | 9 | 704 | |||
CR457041.1 | - | 9 | 638 | |||
DQ411039.1 | - | 8 | 542 | |||
KU177968.1 | - | 9 | 638 | |||
KU177969.1 | - | 8 | 542 | |||
L04658.1 | - | 9 | 650 | |||
X66364.1 | - | 9 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
