siRNA Details
Gene Symbol: CDK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164410.2 | NP_001157882.1 | 3 | 302 | ||
NM_004935.3 | NP_004926.1 | 3 | 302 | |||
GenBank mRNA | AK026533.1 | - | 3 | 223 | ||
AY049778.1 | - | 3 | 185 | |||
BC005115.1 | - | 3 | 210 | 3537202 | ||
BP309068.1 | - | 3 | 282 | |||
BT006680.1 | - | 3 | 185 | |||
BU527089.1 | - | 3 | 250 | |||
CB114175.1 | - | 3 | 228 | |||
CR457041.1 | - | 3 | 185 | |||
DB487008.1 | - | 3 | 302 | |||
DQ411039.1 | - | 3 | 185 | |||
KU177968.1 | - | 3 | 185 | |||
KU177969.1 | - | 3 | 185 | |||
L04658.1 | - | 3 | 197 | |||
X66364.1 | - | 3 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
