siRNA Details
Gene Symbol: SIRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193286.1 | NP_001180215.1 | 13 | 1868 | ||
NM_012237.3 | NP_036369.2 | 16 | 2028 | |||
NM_030593.2 | NP_085096.1 | 15 | 2020 | |||
NR_034146.1 | 14 | 1951 | ||||
XM_006723111.1 | XP_006723174.1 | 17 | 1976 | |||
XM_011526654.1 | XP_011524956.1 | 15 | 1913 | |||
XM_011526655.1 | XP_011524957.1 | 13 | 1728 | |||
XM_017026500.1 | XP_016881989.1 | 15 | 1952 | |||
GenBank mRNA | AB009274.1 | - | 15 | 1748 | ||
AF083107.2 | - | 16 | 1926 | |||
AF095714.1 | - | 15 | 1801 | |||
AF131800.1 | - | 12 | 1521 | |||
AF160214.1 | - | 15 | 1778 | |||
AJ505014.1 | - | 13 | 2498 | |||
AK025876.1 | - | 15 | 1806 | |||
AK054642.1 | - | 16 | 1859 | |||
AK131390.1 | - | 5 | 4693 | |||
AK225824.1 | - | 15 | 1802 | |||
AY030277.1 | - | 15 | 1780 | |||
BC003012.1 | - | 15 | 1871 | 2820929 | ||
BC003547.1 | - | 15 | 1871 | 3536728 | ||
BF529906.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -





