Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271957.1 | NP_001258886.1 | 4 | 1255 | ||
NM_001271958.1 | NP_001258887.1 | 4 | 1092 | |||
NM_001271959.1 | NP_001258888.1 | 4 | 1261 | |||
NM_001271960.1 | NP_001258889.1 | 5 | 1153 | |||
NM_001271961.1 | NP_001258890.1 | 3 | 961 | |||
NM_014437.4 | NP_055252.2 | 5 | 1399 | |||
GenBank mRNA | AF132942.1 | - | 3 | 949 | ||
AF151829.1 | - | 4 | 970 | |||
AJ243649.1 | - | 4 | 986 | |||
AJ271671.1 | - | 4 | 1000 | |||
AK074943.1 | - | 4 | 1209 | |||
AK293389.1 | - | 3 | 869 | |||
AK297983.1 | - | 5 | 967 | |||
AK300244.1 | - | 5 | 959 | |||
BC002563.2 | - | 4 | 974 | 3138575 | ||
BC003152.2 | - | 3 | 946 | 2958651 | ||
BC007886.2 | - | 4 | 971 | 4122086 | ||
BC014303.2 | - | 5 | 1379 | 3942586 |