Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 6 | 1375 | ||
NM_016166.2 | NP_057250.1 | 5 | 776 | |||
XM_011522126.2 | XP_011520428.1 | 7 | 1483 | |||
XM_011522127.2 | XP_011520429.1 | 5 | 1260 | |||
XM_017022688.1 | XP_016878177.1 | 5 | 1254 | |||
XM_017022689.1 | XP_016878178.1 | 5 | 1267 | |||
XM_017022690.1 | XP_016878179.1 | 5 | 1072 | |||
XM_017022691.1 | XP_016878180.1 | 5 | 846 | |||
GenBank mRNA | AF077951.1 | - | 5 | 628 | ||
AF167160.1 | - | 5 | 724 | |||
AK094641.1 | - | 6 | 1375 | |||
AK314515.1 | - | 5 | 651 | |||
AY826819.1 | - | 5 | 625 | |||
BC053685.1 | - | 5 | 629 | |||
BC104646.1 | - | 5 | 644 | |||
BC118587.1 | - | 5 | 678 | 40085010 | ||
BC121797.1 | - | 5 | 675 | 40085007 | ||
EF487997.1 | - | 5 | 628 | |||
EF487998.1 | - | 5 | 628 | |||
FJ997900.1 | - | 5 | 628 | |||
U78524.1 | - | 4 | 610 |