Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018682.3 | NP_061152.3 | 4 | 732 | ||
NM_182931.2 | NP_891847.1 | 5 | 880 | |||
XM_005250493.1 | XP_005250550.1 | 4 | 799 | |||
XM_011516400.1 | XP_011514702.1 | 6 | 1006 | |||
XM_017012435.1 | XP_016867924.1 | 4 | 791 | |||
GenBank mRNA | AF067804.1 | - | 1 | 199 | ||
AF519459.1 | - | 4 | 547 | |||
AK000940.1 | - | 4 | 547 | |||
AK024088.1 | - | 1905 | ||||
AY147037.1 | - | 5 | 880 | |||
AY157990.1 | - | 3 | 346 | |||
BC001296.2 | - | 5 | 842 | |||
BC016382.1 | - | 1 | 45 | |||
BC024658.1 | - | 1 | 175 | |||
BC040004.1 | - | 4 | 750 | |||
BC053906.1 | - | 4 | 788 | |||
BC062583.1 | - | 5 | 852 | |||
BC142987.1 | - | 4 | 762 | |||
JQ809698.1 | - | 3 | 346 |