Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004964.2 | NP_004955.2 | 5 | 457 | ||
XM_011541309.2 | XP_011539611.1 | 5 | 461 | |||
GenBank mRNA | AB451297.1 | - | 5 | 394 | ||
AB451430.1 | - | 5 | 394 | |||
AK225555.1 | - | 5 | 462 | |||
AK296308.1 | - | 4 | 347 | |||
AK299241.1 | - | 5 | 391 | |||
AK299792.1 | - | 5 | 417 | |||
AK313061.1 | - | 5 | 460 | |||
AY627042.1 | - | 5 | 401 | |||
BC000301.2 | - | 5 | 428 | 2820260 | ||
CR541717.1 | - | 5 | 394 | |||
D50405.1 | - | 5 | 457 | |||
U50079.1 | - | 5 | 504 |