Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286752.1 | NP_001273681.1 | 10 | 1689 | ||
NM_017438.4 | NP_059134.1 | 9 | 1546 | |||
NR_040087.2 | 8 | 1457 | ||||
XM_011529636.1 | XP_011527938.1 | 9 | 1595 | |||
XM_011529637.1 | XP_011527939.1 | 9 | 1660 | |||
XM_011529638.2 | XP_011527940.1 | 9 | 1568 | |||
XM_011529639.1 | XP_011527941.1 | 9 | 1346 | |||
XM_011529640.2 | XP_011527942.1 | 9 | 1611 | |||
XM_011529642.1 | XP_011527944.1 | 10 | 1689 | |||
XM_011529643.1 | XP_011527945.1 | 10 | 1640 | |||
XM_011529644.1 | XP_011527946.1 | 10 | 1201 | |||
XM_017028403.1 | XP_016883892.1 | 9 | 1137 | |||
XM_017028404.1 | XP_016883893.1 | 10 | 1245 | |||
GenBank mRNA | AF391112.1 | - | 8 | 1024 | ||
AK001660.1 | - | 9 | 1113 | |||
AK095215.1 | - | 8 | 2413 | |||
AK126368.1 | - | 2 | 1623 | |||
BC036556.2 | - | 10 | 1363 | 5266627 |