siRNA Details
Gene Symbol: IFNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021057.2 | NP_066401.2 | 1 | 185 | ||
GenBank mRNA | BC074991.2 | - | 1 | 183 | 30915521 | |
BC074992.2 | - | 1 | 183 | 30915307 | ||
BC113994.1 | - | 1 | 185 | 40084821 | ||
BC114000.1 | - | 1 | 185 | 40084820 | ||
M34913.1 | - | 1 | 185 |
Gene Symbol: IFNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002171.2 | NP_002162.1 | 1 | 191 | ||
GenBank mRNA | BC069409.1 | - | 1 | 191 | 7262186 | |
BC103972.2 | - | 1 | 191 | 40015081 | ||
BC103973.1 | - | 1 | 191 | 40015082 | ||
V00551.1 | - | 1 | 191 |
Gene Symbol: IFNA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021268.2 | NP_067091.1 | 1 | 194 | ||
GenBank mRNA | BC096732.1 | - | 1 | 187 | 40015059 | |
BC098355.1 | - | 1 | 187 | 40015056 | ||
M11026.1 | - | 1 | 194 | |||
M38289.1 | - | 1 | 287 | |||
M71246.1 | - | 1 | 106 |
Gene Symbol: IFNA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002175.2 | NP_002166.2 | 1 | 193 | ||
GenBank mRNA | BC069329.1 | - | 1 | 153 | 7262150 | |
BC069372.1 | - | 1 | 153 | 7262162 | ||
BC069408.1 | - | 1 | 153 | 7262174 | ||
BC096699.1 | - | 1 | 187 | 40015057 | ||
BC101638.1 | - | 1 | 152 | 8069144 | ||
BC101640.1 | - | 1 | 152 | 8069146 | ||
M12350.1 | - | 1 | 193 | |||
M28586.1 | - | 1 | 193 | |||
V00540.1 | - | 1 | 153 | |||
X00145.1 | - | 1 | 193 |
Gene Symbol: IFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002169.2 | NP_002160.1 | 1 | 202 | ||
GenBank mRNA | BC093755.1 | - | 1 | 202 | 7939600 | |
BC093757.1 | - | 1 | 202 | 7939602 |
Gene Symbol: IFNA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002170.3 | NP_002161.2 | 1 | 175 | ||
GenBank mRNA | BC104828.1 | - | 1 | 174 | 8143831 | |
BC104830.1 | - | 1 | 174 | 8143833 | ||
K01900.1 | - | 1 | 189 | |||
V00545.1 | - | 1 | 176 | |||
V00550.1 | - | 1 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
