siRNA Details
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024013.2 | NP_076918.1 | 1 | 196 | ||
GenBank mRNA | AB578886.1 | - | 1 | 209 | ||
BC074928.2 | - | 1 | 172 | 30915487 | ||
BC074929.2 | - | 1 | 172 | 30915263 | ||
BC112300.1 | - | 1 | 180 | 8327768 | ||
BC112302.1 | - | 1 | 180 | 8327770 | ||
JN848523.1 | - | 1 | 142 | |||
V00537.1 | - | 1 | 198 | |||
V00538.1 | - | 1 | 196 |
Gene Symbol: IFNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006900.3 | NP_008831.3 | 1 | 211 | ||
GenBank mRNA | BC069427.1 | - | 1 | 192 | 7262166 | |
BC093988.1 | - | 1 | 183 | 7939833 | ||
BC112002.1 | - | 1 | 183 | 8327470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
