Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014390.2 | NP_055205.2 | 22 | 2835 | ||
GenBank mRNA | AB209510.1 | - | 22 | 2773 | ||
AK096583.1 | - | 23 | 2760 | |||
AK304181.1 | - | 22 | 2727 | |||
AK309014.1 | - | 14 | 1728 | |||
AK312313.1 | - | 22 | 2789 | |||
BC017180.2 | - | 22 | 2791 | 3345037 | ||
BC020423.1 | - | 1 | 76 | |||
BT009785.1 | - | 22 | 2609 | |||
U22055.1 | - | 22 | 2801 |