Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDPK1 SIRNA Assays ›
          Gene Symbol:
          PDPK1
          Gene Name:
          3-phosphoinositide dependent protein kinase 1
          Gene Aliases:
          PDK1, PDPK2, PDPK2P, PRO0461
          Chromosome Location:
          Chr.16: 2537964 - 2603190 on Build GRCh38
          UniGene ID:
          Hs.459691
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 214313

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDPK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001261816.1 NP_001248745.1 10 1160
          NM_002613.4 NP_002604.1 10 1160
          NM_031268.5 NP_112558.2 7 779
          XM_011522521.2 XP_011520823.1 10 4848
          XM_011522523.2 XP_011520825.1 7 4472
          XM_017023271.1 XP_016878760.1 10 4853
          XM_017023272.1 XP_016878761.1 10 1409
          XM_017023273.1 XP_016878762.1 10 1118
          XM_017023274.1 XP_016878763.1 11 1256
          XM_017023275.1 XP_016878764.1 7 4478
          XM_017023276.1 XP_016878765.1 7 722
          XM_017023277.1 XP_016878766.1 8 820
          GenBank mRNA AB209835.1 - 10 1127
          AF017995.1 - 10 1091
          AJ785968.1 - 9 1794
          AK131060.1 - 10 1094
          AK222581.1 - 10 1103
          BC006339.2 - 7 740 4079551
          BC012103.1 - 10 1120 4561371
          BC033494.1 - 7 692 5176294
          CR536517.1 - 10 1011
          Y15056.1 - 10 1090

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          type B pancreatic cell development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of protein kinase activity
          hyperosmotic response
          epidermal growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          regulation of endothelial cell migration
          negative regulation of cardiac muscle cell apoptotic process
          cell migration
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          calcium-mediated signaling
          actin cytoskeleton organization
          platelet activation
          negative regulation of transforming growth factor beta receptor signaling pathway
          T cell costimulation
          activation of protein kinase B activity
          cellular response to insulin stimulus
          negative regulation of toll-like receptor signaling pathway
          intracellular signal transduction
          Fc-epsilon receptor signaling pathway
          regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of mast cell degranulation
          protein autophosphorylation
          phosphatidylinositol-mediated signaling
          focal adhesion assembly
          T cell receptor signaling pathway
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to epidermal growth factor stimulus
          positive regulation of establishment of protein localization to plasma membrane
          extrinsic apoptotic signaling pathway
          protein serine/threonine kinase activity
          3-phosphoinositide-dependent protein kinase activity
          insulin receptor binding
          protein binding
          ATP binding
          phospholipase activator activity
          kinase activity
          protein kinase binding
          phospholipase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0