Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008859.2 | NP_032885.1 | 9 | 967 | ||
GenBank mRNA | AB062122.1 | - | 2 | 365 | ||
AK037664.1 | - | 9 | 1039 | |||
AK085546.1 | - | 9 | 1237 | |||
BC138552.1 | - | 8 | 898 | 8861572 | ||
BC145769.1 | - | 8 | 898 | 40131072 | ||
D11091.1 | - | 9 | 967 |