Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCTD SIRNA Assays ›
Gene Symbol:
DCTD
Gene Name:
dCMP deaminase
Gene Aliases:
-
Chromosome Location:
Chr.4: 182890091 - 182917477 on Build GRCh38
UniGene ID:
Hs.183850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
214555
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCTD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012732.1
NP_001012750.1
6
684
NM_001921.2
NP_001912.2
6
661
XM_005262778.3
XP_005262835.1
6
709
XM_005262779.3
XP_005262836.1
7
800
XM_005262780.3
XP_005262837.1
8
808
XM_005262781.4
XP_005262838.1
5
466
XM_005262782.3
XP_005262839.1
5
463
XM_006714115.3
XP_006714178.1
6
607
XM_006714116.3
XP_006714179.1
5
904
XM_011531674.2
XP_011529976.1
7
1294
XM_011531675.2
XP_011529977.1
6
1025
XM_011531676.2
XP_011529978.1
7
1033
XM_017007816.1
XP_016863305.1
7
776
XM_017007817.1
XP_016863306.1
7
743
XM_017007818.1
XP_016863307.1
8
808
XM_017007819.1
XP_016863308.1
8
1302
XM_017007820.1
XP_016863309.1
8
1165
XM_017007821.1
XP_016863310.1
8
1298
XM_017007822.1
XP_016863311.1
8
643
XM_017007823.1
XP_016863312.1
5
754
XM_017007824.1
XP_016863313.1
5
381
GenBank mRNA
AK293135.1
-
5
440
AK310788.1
-
5
444
AK313221.1
-
6
586
BC001286.1
-
6
536
3458999
BC088357.1
-
6
575
30389169
BU188090.1
-
5
490
L12136.1
-
5
487
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001161516, NM_178788, NM_001161515, BC031719, CO803572, and others.
Panther Classification:
Molecular Function -
deaminase
hydrolase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleotide metabolic process
nucleotide biosynthetic process
pyrimidine nucleoside biosynthetic process
dCMP deaminase activity
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0