Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADARB1 SIRNA Assays ›
          Gene Symbol:
          ADARB1
          Gene Name:
          adenosine deaminase, RNA specific B1
          Gene Aliases:
          ADAR2, DRABA2, DRADA2, RED1
          Chromosome Location:
          Chr.21: 45074578 - 45226563 on Build GRCh38
          UniGene ID:
          Hs.474018
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 214572

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADARB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001112.3 NP_001103.1 5, 6 1521
          NM_001160230.1 NP_001153702.1 5, 6 1521
          NM_015833.3 NP_056648.1 5, 6 1521
          NM_015834.3 NP_056649.1 5, 6 1521
          NR_027672.1 4, 5 586
          NR_027673.1 6, 7 1639
          NR_027674.1 5, 6 1521
          NR_073200.1 5, 6 1521
          XM_006723954.2 XP_006724017.1 6, 7 1593
          XM_011529425.2 XP_011527727.1 6, 7 2713
          XM_011529432.1 XP_011527734.1 4, 5 434
          XM_017028242.1 XP_016883731.1 4, 5 3189
          XM_017028243.1 XP_016883732.1 7, 8 6678
          XM_017028244.1 XP_016883733.1 4, 5 3187
          XM_017028245.1 XP_016883734.1 4, 5 3142
          XM_017028246.1 XP_016883735.1 7, 8 6678
          XM_017028247.1 XP_016883736.1 6, 7 6560
          XM_017028248.1 XP_016883737.1 7, 8 1502
          XM_017028249.1 XP_016883738.1 8, 9 6721
          XM_017028250.1 XP_016883739.1 7, 8 6631
          XM_017028251.1 XP_016883740.1 4, 5 1303
          XM_017028252.1 XP_016883741.1 4, 5 3189
          XM_017028253.1 XP_016883742.1 4, 5 3137
          XM_017028254.1 XP_016883743.1 6, 7 6560
          XM_017028255.1 XP_016883744.1 6, 7 6558
          XM_017028256.1 XP_016883745.1 5, 6 1353
          GenBank mRNA AB194369.1 - 4, 5 564
          AB194370.1 - 5, 6 1499
          AB194371.1 - 5, 6 1499
          AB194372.2 - 5, 6 1499
          AB194373.1 - 5, 6 1499
          AF001042.1 - 3, 4 1101
          AF525422.1 - 6, 7 1617
          AF533142.1 - 5, 6 1499
          AY135659.1 - 6, 7 1617
          BC065545.1 - 5, 6 1358 6014605
          U76420.1 - 5, 6 1499
          U76421.1 - 5, 6 1499
          U76422.1 - 5, 6 1499
          U82120.1 - 5, 6 1326
          U82121.1 - 3, 4 1086
          X99227.1 - 4, 5 1233
          X99383.1 - 4, 5 1233

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          adenosine to inosine editing
          mRNA processing
          negative regulation of cell proliferation
          negative regulation of cell migration
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          positive regulation of viral genome replication
          innate immune response
          defense response to virus
          regulation of cell cycle
          double-stranded RNA adenosine deaminase activity
          mRNA binding
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0