Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APEX1 SIRNA Assays ›
          Gene Symbol:
          APEX1
          Gene Name:
          apurinic/apyrimidinic endodeoxyribonuclease 1
          Gene Aliases:
          APE, APE1, APEN, APEX, APX, HAP1, REF1
          Chromosome Location:
          Chr.14: 20455131 - 20457772 on Build GRCh38
          UniGene ID:
          Hs.73722
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 214732

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: APEX1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001244249.1 NP_001231178.1 4 625
          NM_001641.3 NP_001632.2 4 630
          NM_080648.2 NP_542379.1 4 564
          NM_080649.2 NP_542380.1 4 574
          GenBank mRNA AK098588.1 - 4 514
          AK291100.1 - 4 539
          BC002338.2 - 4 515 2819505
          BC004979.1 - 4 427 2905681
          BC008145.1 - 4 513 4180383
          BC019291.1 - 4 515 2823545
          BC095428.1 - 5 504 30709297
          BT007236.1 - 3 298
          BT020133.1 - 3 298
          BX161454.1 - 4 464
          D90373.1 - 4 503
          M80261.1 - 4 420
          M81955.1 - 4 508
          S43127.1 - 4 521
          U79268.1 - 4 476
          X59764.1 - 4 507

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          DNA repair
          base-excision repair
          base-excision repair, base-free sugar-phosphate removal
          DNA recombination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          aging
          negative regulation of smooth muscle cell migration
          response to drug
          regulation of mRNA stability
          cell redox homeostasis
          positive regulation of DNA repair
          oxidation-reduction process
          cellular response to hydrogen peroxide
          cellular response to cAMP
          cellular response to peptide hormone stimulus
          DNA demethylation
          RNA phosphodiester bond hydrolysis, endonucleolytic
          positive regulation of G1/S transition of mitotic cell cycle
          negative regulation of nucleic acid-templated transcription
          DNA binding
          damaged DNA binding
          double-stranded telomeric DNA binding
          transcription coactivator activity
          transcription corepressor activity
          DNA-(apurinic or apyrimidinic site) lyase activity
          endonuclease activity
          endodeoxyribonuclease activity
          RNA-DNA hybrid ribonuclease activity
          phosphodiesterase I activity
          uracil DNA N-glycosylase activity
          protein binding
          phosphoric diester hydrolase activity
          double-stranded DNA exodeoxyribonuclease activity
          double-stranded DNA 3'-5' exodeoxyribonuclease activity
          3'-5' exonuclease activity
          oxidoreductase activity
          site-specific endodeoxyribonuclease activity, specific for altered base
          chromatin DNA binding
          protein complex binding
          poly(A) RNA binding
          metal ion binding
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0