Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153209.3 | NP_694941.2 | 12 | 1632 | ||
XM_005257029.1 | XP_005257086.1 | 12 | 1632 | |||
XM_005257030.1 | XP_005257087.1 | 11 | 1506 | |||
XM_017024150.1 | XP_016879639.1 | 12 | 1671 | |||
XM_017024151.1 | XP_016879640.1 | 12 | 1671 | |||
XM_017024152.1 | XP_016879641.1 | 11 | 1545 | |||
XM_017024153.1 | XP_016879642.1 | 11 | 1545 | |||
XM_017024154.1 | XP_016879643.1 | 11 | 1506 | |||
XM_017024155.1 | XP_016879644.1 | 11 | 1485 | |||
XM_017024156.1 | XP_016879645.1 | 11 | 1485 | |||
XM_017024157.1 | XP_016879646.1 | 10 | 1359 | |||
XM_017024158.1 | XP_016879647.1 | 10 | 1359 | |||
GenBank mRNA | AK094619.1 | - | 12 | 1587 | ||
BC026362.1 | - | 11 | 1486 | |||
BC093631.1 | - | 12 | 1518 | 7939476 | ||
BC104852.1 | - | 12 | 1518 | 8143855 | ||
BC110989.1 | - | 12 | 1596 | |||
BC143327.1 | - | 11 | 1393 | 9051832 |