Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT10 SIRNA Assays ›
Gene Symbol:
SEPT10
Gene Name:
septin 10
Gene Aliases:
-
Chromosome Location:
Chr.2: 109542797 - 109614247 on Build GRCh38
UniGene ID:
Hs.469615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215107
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321496.1
NP_001308425.1
3, 4
661
NM_001321498.1
NP_001308427.1
5, 6
975
NM_001321500.1
NP_001308429.1
4, 5
906
NM_001321501.1
NP_001308430.1
5, 6
979
NM_001321502.1
NP_001308431.1
4, 5
906
NM_001321504.1
NP_001308433.1
2, 3
592
NM_001321505.1
NP_001308434.1
2, 3
592
NM_001321507.1
NP_001308436.1
3, 4
661
NM_001321508.1
NP_001308437.1
2, 3
592
NM_001321509.1
NP_001308438.1
5, 6
979
NM_001321512.1
NP_001308441.1
5, 6
975
NM_001321513.1
NP_001308442.1
5, 6
1112
NM_001321514.1
NP_001308443.1
5, 6
803
NM_001321515.1
NP_001308444.1
4, 5
1043
NM_144710.4
NP_653311.1
5, 6
975
NM_178584.3
NP_848699.1
4, 5
906
XM_006712317.1
XP_006712380.1
5, 6
975
XM_011510698.2
XP_011509000.1
5, 6
808
XM_011510699.2
XP_011509001.1
4, 5
627
XM_011510700.2
XP_011509002.1
5, 6
978
XM_011510701.2
XP_011509003.1
3, 4
702
XM_011510703.2
XP_011509005.1
5, 6
585
XM_011510704.2
XP_011509006.1
5, 6
975
GenBank mRNA
AB208875.1
-
1, 2
69
AF146760.1
-
5, 6
722
AK021681.1
-
5, 6
975
AK022682.1
-
2, 3
307
AK092033.1
-
4, 5
626
AK292038.1
-
5, 6
693
AK295560.1
-
3, 4
371
AL353938.1
-
1, 2
70
BC020502.1
-
5, 6
669
3913356
BC050345.1
-
5, 6
912
4830029
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
cell division
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0