Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003592.2 | NP_003583.2 | 22 | 2827 | ||
XM_005250060.4 | XP_005250117.1 | 22 | 2863 | |||
XM_011516629.2 | XP_011514931.1 | 22 | 2819 | |||
XM_011516630.2 | XP_011514932.1 | 22 | 2720 | |||
XM_011516631.2 | XP_011514933.1 | 22 | 2848 | |||
XM_011516632.2 | XP_011514934.1 | 22 | 2746 | |||
XM_017012723.1 | XP_016868212.1 | 21 | 2583 | |||
GenBank mRNA | AK096163.1 | - | 22 | 2632 | ||
AK096505.1 | - | 15 | 2484 | |||
AK296909.1 | - | 21 | 2552 | |||
BC125119.1 | - | 22 | 2740 | 40118923 | ||
BC125120.1 | - | 22 | 2740 | 40118927 | ||
BX537409.1 | - | 22 | 2757 |