Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130841.1 | NP_001124313.1 | 12 | 1972 | ||
NM_003729.3 | NP_003720.1 | 11 | 1933 | |||
XM_005271298.4 | XP_005271355.1 | 11 | 1930 | |||
XM_005271299.4 | XP_005271356.1 | 11 | 1919 | |||
XM_017002669.1 | XP_016858158.1 | 11 | 1676 | |||
GenBank mRNA | CA437545.1 | - | 1 | 189 |
Gene Symbol: CLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025306.1 | - | 3 | 2181 |
Gene Symbol: DCLRE1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033855.2 | NP_001029027.1 | 14 | 5105 | ||
NM_001033857.2 | NP_001029029.1 | 15 | 5190 | |||
NM_001033858.2 | NP_001029030.1 | 16 | 5512 | |||
NM_001289076.1 | NP_001276005.1 | 12 | 4964 | |||
NM_001289077.1 | NP_001276006.1 | 15 | 5236 | |||
NM_001289078.1 | NP_001276007.1 | 12 | 4997 | |||
NM_001289079.1 | NP_001276008.1 | 16 | 5558 | |||
NM_022487.3 | NP_071932.2 | 13 | 5049 | |||
NR_110297.1 | 17 | 5458 |
Gene Symbol: DSTNP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_033796.2 | 1 | 867 | |||
GenBank mRNA | U72518.1 | - | LIT2102 destrin-2 pseudogene |
1 | 867 |
Gene Symbol: NME7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209049.1 | - | 7 | 1314 |
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW080436.1 | - | 1 | 347 |
Gene Symbol: SMYD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003163.1 | XP_016858652.1 | 1 | 135 |
Gene Symbol: TACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124297.1 | - | 4 | 1171 |
Gene Symbol: THUMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF380578.1 | - | 1 | 494 |
Gene Symbol: TRAPPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011658.3 | NP_001011658.1 | 6 | 1139 | ||
NM_001128835.2 | NP_001122307.2 | 6 | 1133 | |||
NM_014563.5 | NP_055378.1 | 5 | 997 | |||
XM_011545565.1 | XP_011543867.1 | 7 | 1207 | |||
XM_011545566.2 | XP_011543868.1 | 6 | 1063 | |||
GenBank mRNA | AK055671.1 | - | 1 | 304 | ||
BC016915.1 | - | 5 | 960 | 4432196 | ||
BC052618.1 | - | 5 | 960 | 6581382 | ||
BG033605.1 | - | 139 |
Gene Symbol: ZYG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004339.2 | NP_001004339.2 | 14 | 3707 | ||
NM_001307931.1 | NP_001294860.1 | 13 | 2955 | |||
XM_011541480.2 | XP_011539782.1 | 13 | 3437 | |||
XM_011541481.2 | XP_011539783.1 | 13 | 3305 | |||
XM_011541482.2 | XP_011539784.1 | 13 | 3287 | |||
XM_011541483.2 | XP_011539785.1 | 12 | 3242 |