Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003521.2 | NP_003512.1 | 1 | 36 | ||
GenBank mRNA | BC066244.1 | - | 1 | 36 | 7002058 | |
BC067485.1 | - | 1 | 36 | |||
BC067486.1 | - | 1 | 36 | 7002060 | ||
BC067487.1 | - | 1 | 36 | 7002064 | ||
BC067488.1 | - | 1 | 36 | 7002065 | ||
BC067489.1 | - | 1 | 36 | 7002066 |