Search
Search

CMTM4| Gene Symbol: CMTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178818.2 | NP_848933.1 | 1, 2 | 364 | ||
| NM_181521.2 | NP_852662.1 | 1, 2 | 364 | |||
| XM_017022954.1 | XP_016878443.1 | 1, 2 | 268 | |||
| GenBank mRNA | AF479814.1 | - | 1, 2 | 364 | ||
| AF521889.1 | - | 1, 2 | 364 | |||
| BC093679.1 | - | 1, 2 | 246 | 7939524 | ||