Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS1 SIRNA Assays ›
Gene Symbol:
HPS1
Gene Name:
HPS1, biogenesis of lysosomal organelles complex 3 subunit 1
Gene Aliases:
BLOC3S1, HPS
Chromosome Location:
Chr.10: 98415068 - 98446963 on Build GRCh38
UniGene ID:
Hs.404568
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215735
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000195.4
NP_000186.2
3
306
NM_001311345.1
NP_001298274.1
3
315
NM_001322476.1
NP_001309405.1
3
235
NM_001322477.1
NP_001309406.1
3
315
NM_001322478.1
NP_001309407.1
3
235
NM_001322479.1
NP_001309408.1
3
315
NM_001322480.1
NP_001309409.1
3
235
NM_001322481.1
NP_001309410.1
3
315
NM_001322482.1
NP_001309411.1
3
315
NM_001322483.1
NP_001309412.1
3
235
NM_001322484.1
NP_001309413.1
3
315
NM_001322485.1
NP_001309414.1
3
306
NM_001322487.1
NP_001309416.1
3
315
NM_001322489.1
NP_001309418.1
3
315
NM_001322490.1
NP_001309419.1
3
315
NM_001322491.1
NP_001309420.1
3
235
NM_001322492.1
NP_001309421.1
3
235
NM_182639.3
NP_872577.1
3
315
XM_005269757.4
XP_005269814.1
3
248
XM_017016170.1
XP_016871659.1
3
222
XM_017016171.1
XP_016871660.1
3
311
XM_017016172.1
XP_016871661.1
3
283
XM_017016173.1
XP_016871662.1
3
313
GenBank mRNA
AK295821.1
-
3
271
AL702912.1
-
3
300
BC000175.2
-
3
265
BI518436.1
-
3
225
DA385131.1
-
3
313
DA479757.1
-
3
235
KU178114.1
-
1
44
KU178115.1
-
1
44
KU178116.1
-
1
44
KU178117.1
-
1
44
KU178118.1
-
1
44
KU178119.1
-
1
44
KU178120.1
-
1
44
KU178121.1
-
1
44
KU178122.1
-
1
44
U65676.1
-
3
250
U96721.1
-
3
278
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019424, AK154994, BG062886, BC089331, BC058332, and others.
Gene Ontology Categories:
Function(s)
Process(es)
lysosome organization
visual perception
response to stimulus
protein binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-7wptj:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0