Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 22 | 4090 | ||
NM_022489.3 | NP_071934.3 | 23 | 4147 | |||
XM_005268004.4 | XP_005268061.1 | 23 | 4234 | |||
XM_005268005.4 | XP_005268062.1 | 22 | 4148 | |||
GenBank mRNA | AF318379.1 | - | 5 | 1231 | ||
AK025709.1 | - | 14 | 2110 | |||
AK027105.1 | - | 2 | 543 | |||
AL832905.1 | - | 2 | 568 | |||
BC008756.2 | - | 12 | 1998 | |||
BC064828.1 | - | 7 | 1465 | |||
DQ395338.1 | - | 15 | 2351 | |||
DQ395339.1 | - | 16 | 2408 | |||
DQ395340.1 | - | 17 | 2492 |