Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@2c2deaa7
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
See other TCF7 SIRNA Assays ›
Gene Symbol:
TCF7
Gene Name:
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases:
TCF-1
Chromosome Location:
Chr.5: 134114703 - 134148229 on Build GRCh38
UniGene ID:
Hs.573153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215766
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price (USD)
233.00
Your Price (USD)
Online offer (USD):
233.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134851.2
NP_001128323.2
3, 4
296
NM_003202.3
NP_003193.2
4, 5
740
NM_201632.3
NP_963963.1
3, 4
296
NM_201634.3
NP_963965.1
3, 4
296
NM_213648.3
NP_998813.1
2, 3
530
NR_033449.1
2, 3
531
XM_006714678.3
XP_006714741.1
4, 5
748
XM_006714679.3
XP_006714742.1
4, 5
748
XM_006714680.3
XP_006714743.1
4, 5
748
XM_006714682.2
XP_006714745.1
4, 5
748
XM_006714684.2
XP_006714747.1
4, 5
748
XM_006714685.3
XP_006714748.1
3, 4
293
XM_006714686.3
XP_006714749.1
3, 4
295
XM_006714687.3
XP_006714750.1
2, 3
160
XM_011543604.2
XP_011541906.1
4, 5
748
XM_011543606.1
XP_011541908.1
4, 5
748
XM_011543607.2
XP_011541909.1
2, 3
6236
XM_011543608.2
XP_011541910.1
2, 3
2281
XM_011543609.2
XP_011541911.1
2, 3
242
XM_011543610.2
XP_011541912.1
2, 3
281
XM_011543611.2
XP_011541913.1
2, 3
1175
XM_011543612.2
XP_011541914.1
2, 3
242
XM_011543613.2
XP_011541915.1
2, 3
281
XM_017009789.1
XP_016865278.1
3, 4
293
XM_017009790.1
XP_016865279.1
2, 3
1175
GenBank mRNA
AK057580.1
-
2, 3
531
AK093530.1
-
2, 3
401
AK310591.1
-
2, 3
242
AL832816.1
-
4, 5
456
AL834166.1
-
3, 4
264
BC048769.1
-
4, 5
740
6063270
BC072023.1
-
4, 5
675
6292932
DA627229.1
-
3, 4
296
DB145426.1
-
3, 4
296
X59869.1
-
3, 4
278
X59870.1
-
3, 4
278
X59871.1
-
3, 4
278
Z47361.1
-
2, 3
200
Z47362.1
-
2, 3
200
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-1-prod-green:8080/10.253.225.215:8080.
git-commit: 0f6c802f6da5e59cd1b986a4dc444b4bcf3ac46d
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.15.0-2021.01.02-1.0