Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178439.4 | NP_848526.1 | 5 | 956 | ||
XM_011533033.1 | XP_011531335.1 | 5 | 920 | |||
XM_011533034.2 | XP_011531336.1 | 5 | 963 | |||
XM_017004704.1 | XP_016860193.1 | 6 | 985 | |||
XM_017004705.1 | XP_016860194.1 | 3 | 230 | |||
GenBank mRNA | AK023119.1 | - | 5 | 862 | ||
AK023261.1 | - | 4 | 298 | |||
AK024042.1 | - | 5 | 920 | |||
BC007420.2 | - | 5 | 876 | 3030159 |
Gene Symbol: GMCL1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF198534.1 | - | 1 | 252 |