Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GON7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032490.4 | NP_115879.2 | 2 | 453 | ||
GenBank mRNA | AF277185.1 | - | 2 | 198 | ||
BC021733.2 | - | 2 | 433 | 4796574 | ||
BC065216.1 | - | 2 | 355 | |||
BC107429.1 | - | 2 | 421 | 4809642 | ||
BC109390.1 | - | 2 | 364 | |||
BF131439.1 | - | 2 | 455 |