Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144994.7 | NP_659431.5 | 7 | 742 | ||
GenBank mRNA | AF492401.1 | - | 7 | 718 | ||
AJ315766.1 | - | 6 | 686 | |||
AK092564.1 | - | 6 | 616 | |||
AY196212.1 | - | 7 | 726 | |||
BC107056.2 | - | 7 | 737 | 40005436 |