Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000101.3 | NP_000092.2 | 4 | 336 | ||
XM_011522905.2 | XP_011521207.1 | 4 | 331 | |||
GenBank mRNA | AK097127.1 | - | 4 | 239 | ||
AK300049.1 | - | 4 | 240 | |||
AK309030.1 | - | 4 | 240 | |||
BC006465.1 | - | 4 | 292 | 3502788 | ||
BT006861.1 | - | 4 | 265 | |||
BX109994.1 | - | 4 | 301 | |||
M21186.1 | - | 4 | 293 |