Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR9LP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026850.1 | 6 | 1933 | |||
GenBank mRNA | AK055560.1 | - | 7 | 1665 | ||
AK096109.1 | - | 5 | 1485 | |||
AL833686.1 | - | 8 | 2233 | |||
BC016171.1 | - | 3 | 778 | |||
BC041632.1 | - | 3 | 588 |