Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO80C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098817.1 | NP_001092287.1 | 5, 6 | 602 | ||
NM_001308064.1 | NP_001294993.1 | 3, 4 | 365 | |||
NM_194281.3 | NP_919257.2 | 3, 4 | 494 | |||
XM_005258196.4 | XP_005258253.1 | 3, 4 | 312 | |||
XM_005258197.4 | XP_005258254.1 | 3, 4 | 525 | |||
XM_011525812.2 | XP_011524114.1 | 3, 4 | 360 | |||
XM_017025549.1 | XP_016881038.1 | 3, 4 | 332 | |||
GenBank mRNA | AK095502.1 | - | 3, 4 | 436 | ||
AK300660.1 | - | 5, 6 | 554 | |||
BC039404.1 | - | 3, 4 | 460 | 5300795 | ||
BC048989.1 | - | 4, 5 | 422 | |||
BM758571.1 | - | 3, 4 | 267 | |||
BQ441485.1 | - | 3, 4 | 368 | |||
BQ887287.1 | - | 3, 4 | 494 | |||
BY800049.1 | - | 5, 6 | 517 |