Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145450.1 | NP_001138922.1 | 5 | 543 | ||
GenBank mRNA | AY367766.1 | - | 5 | 543 | ||
BC095472.1 | - | 3 | 353 | 30561658 | ||
BC102032.1 | - | 5 | 440 | 40033539 | ||
BC102034.1 | - | 5 | 440 | 40033540 | ||
BC102035.1 | - | 5 | 440 | 40033541 |