Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1LC3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004343.2 | NP_001004343.1 | 4 | 1051 | ||
XM_005273139.3 | XP_005273196.1 | 5 | 1079 | |||
GenBank mRNA | AF276659.1 | - | 4 | 1040 |