Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033393.2 | NP_203751.2 | 6 | 953 | ||
XM_005263319.3 | XP_005263376.1 | 7 | 1094 | |||
XM_006714393.3 | XP_006714456.1 | 7 | 1103 | |||
XM_011532388.1 | XP_011530690.1 | 9 | 1585 | |||
XM_011532389.1 | XP_011530691.1 | 7 | 1699 | |||
XM_017008787.1 | XP_016864276.1 | 5 | 938 | |||
GenBank mRNA | AB051514.1 | - | 6 | 840 |